Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suz12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003529.2 | NP_001003529.1 | 2 - 3 | 445 | 151 | |
XM_021471664.1 | XP_021327339.1 | 2 - 3 | 1460 | 151 | ||
GenBank mRNA | BC078293.1 | - | 2 - 3 | 448 | 151 | |
BC152625.1 | - | 2 - 3 | 334 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |