Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003573.2 | NP_001003573.1 | 5 - 6 | 619 | 71 | |
GenBank mRNA | BC078202.1 | - | 5 - 6 | 623 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |