Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glrbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003587.1 | NP_001003587.1 | 8 - 9 | 1197 | 85 | |
XM_021481003.1 | XP_021336678.1 | 8 - 9 | 1210 | 85 | ||
GenBank mRNA | AB195560.1 | - | 7 - 8 | 897 | 85 | |
BC078269.1 | - | 8 - 9 | 1197 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |