Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glrbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003587.1 | NP_001003587.1 | 9 - 10 | 1487 | 133 | |
XM_005157317.4 | XP_005157374.1 | 2 - 3 | 927 | 133 | ||
XM_021481003.1 | XP_021336678.1 | 9 - 10 | 1500 | 133 | ||
GenBank mRNA | AB195560.1 | - | 8 - 9 | 1187 | 133 | |
BC078269.1 | - | 9 - 10 | 1487 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |