Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111195.1 | NP_001104665.1 | 5 - 6 | 386 | 91 | |
GenBank mRNA | BC079502.1 | - | 5 - 6 | 400 | 91 | |
BC155162.1 | - | 5 - 6 | 386 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |