Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003831.1 | NP_001003831.1 | 701 | 62 | ||
GenBank mRNA | AY648711.1 | - | 701 | 62 | ||
BC085643.1 | - | 694 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio |