Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003832.1 | NP_001003832.1 | 3 - 4 | 222 | 79 | |
GenBank mRNA | AY648827.1 | - | 3 - 4 | 222 | 79 | |
BC083283.1 | - | 3 - 4 | 232 | 79 | ||
BC152260.1 | - | 3 - 4 | 232 | 79 | ||
BC155769.1 | - | 3 - 4 | 229 | 79 | ||
BC165828.1 | - | 3 - 4 | 233 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |