Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003890.3 | NP_001003890.3 | 5 - 6 | 1285 | 73 | |
GenBank mRNA | AY648752.1 | - | 5 - 6 | 1244 | 73 | |
BC098545.1 | - | 5 - 6 | 1285 | 73 | ||
HM210100.1 | - | 5 - 6 | 1001 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |