Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bet1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003998.2 | NP_001003998.1 | 3 - 4 | 399 | 80 | |
GenBank mRNA | BC080213.1 | - | 3 - 4 | 296 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |