Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fth1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004562.1 | NP_001004562.1 | 4 - 5 | 602 | 74 | |
GenBank mRNA | BC081630.1 | - | 4 - 5 | 602 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |