Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004577.1 | NP_001004577.1 | 6 - 7 | 1383 | 77 | |
GenBank mRNA | BC081650.1 | - | 6 - 7 | 1383 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |