Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY570233.1 | - | 4 - 5 | 853 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |