Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005934.2 | NP_001005934.1 | 1 - 2 | 151 | 68 | |
GenBank mRNA | BC083522.1 | - | 1 - 2 | 131 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |