Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005963.2 | NP_001005963.2 | 1 - 2 | 265 | 100 | |
GenBank mRNA | BC083304.1 | - | 1 - 2 | 208 | 100 | |
BC165360.1 | - | 1 - 2 | 230 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |