Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006046.1 | NP_001006046.1 | 2 - 3 | 110 | 77 | |
GenBank mRNA | BC083362.1 | - | 2 - 3 | 110 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |