Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006046.1 | NP_001006046.1 | 3 - 4 | 168 | 66 | |
GenBank mRNA | BC083362.1 | - | 3 - 4 | 168 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |