Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006081.1 | NP_001006081.1 | 2 - 3 | 239 | 73 | |
GenBank mRNA | BC083239.1 | - | 2 - 3 | 239 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |