Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: borcs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007052.2 | NP_001007053.1 | 3 - 4 | 273 | 106 | |
XM_005163008.4 | XP_005163065.1 | 3 - 4 | 106 | |||
GenBank mRNA | BC083541.1 | - | 3 - 4 | 274 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |