Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007202.1 | NP_001007203.1 | 770 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Danio rerio |