Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fosb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328202.1 | NP_001315131.1 | 3 - 4 | 774 | 105 | |
XM_005173641.4 | XP_005173698.1 | 3 - 4 | 1683 | 105 | ||
XM_005173642.4 | XP_005173699.1 | 3 - 4 | 1682 | 105 | ||
GenBank mRNA | BC085664.1 | - | 3 - 4 | 747 | 105 | |
EH434631.1 | - | 3 - 4 | 784 | 104 | ||
EH460635.1 | - | 3 - 4 | 754 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |