Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4ebp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007354.2 | NP_001007355.1 | 1 - 2 | 191 | 75 | |
GenBank mRNA | BC085595.1 | - | 1 - 2 | 192 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |