Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008628.1 | NP_001008628.1 | 5 - 6 | 1198 | 75 | |
GenBank mRNA | BC086729.1 | - | 5 - 6 | 1198 | 75 | |
BC165767.1 | - | 5 - 6 | 1120 | 75 | ||
BC169189.1 | - | 1181 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |