Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008645.1 | NP_001008645.1 | 7 - 8 | 1341 | 68 | |
GenBank mRNA | BC086711.1 | - | 7 - 8 | 1341 | 68 | |
BC165211.1 | - | 7 - 8 | 1256 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |