Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012501.2 | NP_001012519.2 | 11 - 12 | 1113 | 95 | |
GenBank mRNA | BC076404.1 | - | 8 - 9 | 2953 | 95 | |
BC090184.1 | - | 11 - 12 | 1113 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |