Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mybpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013511.2 | NP_001013529.2 | 8 - 9 | 530 | 76 | |
XM_017353828.2 | XP_017209317.1 | 7 - 8 | 595 | 76 | ||
XM_021470159.1 | XP_021325834.1 | 8 - 9 | 564 | 76 | ||
XM_021470160.1 | XP_021325835.1 | 7 - 8 | 592 | 76 | ||
XM_021470161.1 | XP_021325836.1 | 7 - 8 | 542 | 76 | ||
XM_021470162.1 | XP_021325837.1 | 7 - 8 | 479 | 76 | ||
GenBank mRNA | BC091662.1 | - | 524 | 76 | ||
EB790963.1 | - | 4 - 5 | 297 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |