Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013573.1 | NP_001013591.1 | 6 - 7 | 591 | 151 | |
XM_021481013.1 | XP_021336688.1 | 6 - 7 | 618 | 151 | ||
XM_021481015.1 | XP_021336690.1 | 6 - 7 | 618 | 151 | ||
XM_021481016.1 | XP_021336691.1 | 6 - 7 | 640 | 151 | ||
GenBank mRNA | BC090463.1 | - | 6 - 7 | 591 | 151 | |
BC154362.1 | - | 6 - 7 | 612 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |