Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014338.1 | NP_001014360.1 | 8 - 9 | 1271 | 94 | |
XM_009294917.3 | XP_009293192.1 | 8 - 9 | 1368 | 94 | ||
GenBank mRNA | BC091833.1 | - | 8 - 9 | 1271 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |