Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014371.1 | NP_001014393.1 | 11 - 12 | 2303 | 113 | |
XM_005170889.4 | XP_005170946.1 | 11 - 12 | 2382 | 113 | ||
XM_005170890.4 | XP_005170947.1 | 7 - 8 | 1475 | 113 | ||
GenBank mRNA | BC091789.1 | - | 11 - 12 | 2303 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |