Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chodl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017578.2 | NP_001017578.1 | 2 - 3 | 478 | 66 | |
GenBank mRNA | BC092727.1 | - | 2 - 3 | 480 | 66 | |
BC164239.1 | - | 2 - 3 | 444 | 66 | ||
JQ790525.1 | - | 2 - 3 | 550 | 66 | ||
KM655623.1 | - | 2 - 3 | 410 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |