Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017591.1 | NP_001017591.1 | 3 - 4 | 457 | 73 | |
GenBank mRNA | BC092863.1 | - | 3 - 4 | 457 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |