Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ecrg4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017697.1 | NP_001017697.1 | 3 - 4 | 309 | 76 | |
GenBank mRNA | BC093311.1 | - | 3 - 4 | 309 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |