Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017758.1 | NP_001017758.1 | 2 - 3 | 192 | 83 | |
XM_021470629.1 | XP_021326304.1 | 2 - 3 | 250 | 83 | ||
GenBank mRNA | BC093357.1 | - | 2 - 3 | 192 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |