Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017778.1 | NP_001017778.1 | 5 - 6 | 707 | 78 | |
GenBank mRNA | BC092901.1 | - | 5 - 6 | 707 | 78 | |
BC164579.1 | - | 4 - 5 | 663 | 78 | ||
HM239643.1 | - | 4 - 5 | 551 | 78 | ||
HM239644.1 | - | 4 - 5 | 551 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |