Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017846.1 | NP_001017846.1 | 4 - 5 | 421 | 106 | |
GenBank mRNA | BC092767.1 | - | 4 - 5 | 421 | 106 | |
BC164299.1 | - | 4 - 5 | 416 | 106 | ||
BC171682.1 | - | 4 - 5 | 407 | 106 | ||
BC171686.1 | - | 4 - 5 | 407 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |