Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shisa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017869.1 | NP_001017869.1 | 1 - 2 | 241 | 76 | |
GenBank mRNA | BC092979.1 | - | 1 - 2 | 241 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |