Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: capn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018227.1 | NP_001018177.1 | 20 - 21 | 2135 | 116 | |
GenBank mRNA | BC076522.1 | - | 21 - 22 | 2109 | 116 | |
BC095045.1 | - | 20 - 21 | 2135 | 116 | ||
BC155147.1 | - | 20 - 21 | 2086 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |