Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: capn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018227.1 | NP_001018177.1 | 9 - 10 | 1202 | 135 | |
GenBank mRNA | BC076522.1 | - | 10 - 11 | 1176 | 135 | |
BC095045.1 | - | 9 - 10 | 1202 | 135 | ||
BC155147.1 | - | 9 - 10 | 1153 | 135 | ||
BC165647.1 | - | 9 - 10 | 1164 | 135 | ||
BC171423.1 | - | 9 - 10 | 1153 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |