Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020551.2 | NP_001018387.1 | 3 - 4 | 409 | 98 | |
NM_001045228.2 | NP_001038693.1 | 3 - 4 | 409 | 98 | ||
GenBank mRNA | BC095120.1 | - | 3 - 4 | 403 | 98 | |
BC134979.1 | - | 3 - 4 | 374 | 98 | ||
BC164068.1 | - | 2 - 3 | 334 | 98 | ||
EH531855.1 | - | 5 - 6 | 667 | 98 | ||
EH593412.1 | - | 3 - 4 | 432 | 98 | ||
EH593686.1 | - | 3 - 4 | 407 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |