Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020614.1 | NP_001018450.1 | 4 - 5 | 810 | 128 | |
GenBank mRNA | BC095343.1 | - | 4 - 5 | 810 | 128 | |
BC153552.1 | - | 4 - 5 | 680 | 128 | ||
BC164140.1 | - | 4 - 5 | 575 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |