Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020634.1 | NP_001018470.1 | 672 | 73 | ||
GenBank mRNA | BC095619.1 | - | 672 | 73 | ||
BC159234.1 | - | 737 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio |