Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020754.2 | NP_001018590.2 | 2 - 3 | 424 | 152 | |
GenBank mRNA | BC095375.1 | - | 2 - 3 | 198 | 152 | |
BC155808.1 | - | 2 - 3 | 424 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |