Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020760.1 | NP_001018596.1 | 6 - 7 | 779 | 69 | |
XM_005161708.4 | XP_005161765.1 | 6 - 7 | 661 | 69 | ||
GenBank mRNA | BC095639.1 | - | 6 - 7 | 779 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |