Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024388.1 | NP_001019559.1 | 8 - 9 | 620 | 81 | |
XM_005157569.4 | XP_005157626.1 | 8 - 9 | 603 | 81 | ||
GenBank mRNA | BC095373.1 | - | 8 - 9 | 620 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |