Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024412.1 | NP_001019583.1 | 3 - 4 | 314 | 125 | |
GenBank mRNA | BC095657.1 | - | 3 - 4 | 314 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |