Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024415.2 | NP_001019586.1 | 3 - 4 | 249 | 103 | |
GenBank mRNA | BC095631.1 | - | 3 - 4 | 251 | 103 | |
BC154355.1 | - | 3 - 4 | 248 | 103 | ||
BC164758.1 | - | 3 - 4 | 216 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |