Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320148.1 | NP_001307077.1 | 12 - 13 | 2084 | 101 | |
XM_017354091.2 | XP_017209580.1 | 12 - 13 | 2127 | 101 | ||
GenBank mRNA | BC146747.1 | - | 12 - 13 | 2080 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |