Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099259.1 | NP_001092729.1 | 6 - 7 | 588 | 111 | |
GenBank mRNA | BC142846.1 | - | 6 - 7 | 588 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |