Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105702.1 | NP_001099172.1 | 3 - 4 | 491 | 124 | |
GenBank mRNA | BC148172.1 | - | 3 - 4 | 491 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |