Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002140.1 | NP_001002140.1 | 11 - 12 | 1024 | 78 | |
NM_001144056.1 | NP_001137528.1 | 8 - 9 | 654 | 78 | ||
XM_009307279.3 | XP_009305554.1 | 11 - 12 | 994 | 78 | ||
XM_021480487.1 | XP_021336162.1 | 11 - 12 | 982 | 78 | ||
GenBank mRNA | BC071417.1 | - | 11 - 12 | 1024 | 78 | |
BC095071.1 | - | 10 - 11 | 1067 | 78 | ||
BC165080.1 | - | 8 - 9 | 651 | 78 | ||
BC171457.1 | - | 8 - 9 | 654 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |