Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapre3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002170.1 | NP_001002170.1 | 5 - 6 | 540 | 129 | |
XM_021474991.1 | XP_021330666.1 | 4 - 5 | 550 | 129 | ||
GenBank mRNA | BC074053.1 | - | 5 - 6 | 540 | 129 | |
BC164598.1 | - | 3 - 4 | 455 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |